Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5RAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278167.1 | NP_001265096.1 | 5 | 641 | ||
NM_001278168.1 | NP_001265097.1 | 5 | 641 | |||
NM_001278169.1 | NP_001265098.1 | 5 | 409 | |||
NM_016082.4 | NP_057166.4 | 5 | 493 | |||
NM_016408.3 | NP_057492.2 | 5 | 641 | |||
XM_011528856.2 | XP_011527158.1 | 2 | 1377 | |||
XM_011528857.2 | XP_011527159.1 | 4 | 253 | |||
XM_011528858.1 | XP_011527160.1 | 4 | 492 | |||
XM_011528859.2 | XP_011527161.1 | 4 | 252 | |||
XM_011528860.1 | XP_011527162.1 | 3 | 163 | |||
XM_017027876.1 | XP_016883365.1 | 5 | 602 | |||
GenBank mRNA | AF152097.1 | - | 5 | 577 | ||
AF161516.1 | - | 5 | 592 | |||
AK023992.1 | - | 5 | 591 | |||
AK027449.1 | - | 5 | 441 | |||
AK222745.1 | - | 5 | 591 | |||
AK292075.1 | - | 5 | 359 | |||
AK295342.1 | - | 5 | 577 | |||
AK309906.1 | - | 5 | 591 | |||
AK314532.1 | - | 5 | 591 | |||
AY462283.1 | - | 5 | 592 | |||
AY462284.1 | - | 5 | 592 | |||
BC001215.1 | - | 5 | 570 | 3535564 | ||
BC050706.1 | - | 5 | 584 | 4418659 | ||
BM834342.1 | - | 5 | 444 | |||
DA565688.1 | - | 5 | 443 | |||
DR156307.1 | - | 4 | 250 | |||
KU178644.1 | - | 4 | 479 | |||
KU178645.1 | - | 4 | 479 |