Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016426.6 | NP_057510.4 | 7 | 1571 | ||
XM_005261627.3 | XP_005261684.1 | 7 | 2431 | |||
XM_011530211.2 | XP_011528513.1 | 6 | 2373 | |||
XM_011530213.2 | XP_011528515.1 | 7 | 2433 | |||
XM_017028815.1 | XP_016884304.1 | 6 | 1821 | |||
XM_017028816.1 | XP_016884305.1 | 7 | 2433 | |||
GenBank mRNA | AF223408.1 | - | 6 | 1372 | ||
AK025563.1 | - | 7 | 1398 | |||
AK222806.1 | - | 7 | 1398 | |||
AK303087.1 | - | 6 | 1342 | |||
AK315550.1 | - | 7 | 1397 | |||
AY927426.1 | - | 1 | 311 | |||
AY927427.1 | - | 1 | 311 | |||
BC006325.2 | - | 6 | 1349 | 4138532 | ||
BC075828.1 | - | 7 | 1472 | 5092448 | ||
CT841519.1 | - | 6 | 1315 |