Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003679.4 | NP_003670.2 | 2 | 408 | ||
XM_005273337.1 | XP_005273394.1 | 2 | 518 | |||
XM_005273338.3 | XP_005273395.1 | 3 | 761 | |||
XM_017002649.1 | XP_016858138.1 | 3 | 752 | |||
GenBank mRNA | AF056032.1 | - | 2 | 143 | ||
AK291558.1 | - | 2 | 164 | |||
AK310937.1 | - | 2 | 285 | |||
AU137977.1 | - | 2 | 408 | |||
BC005297.1 | - | 2 | 192 | 3934714 | ||
BP276365.1 | - | 2 | 162 | |||
Y13153.1 | - | 2 | 149 |