Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016441.2 | NP_057525.1 | 6 | 1077 | ||
XM_005264357.4 | XP_005264414.1 | 5 | 899 | |||
XM_011532898.2 | XP_011531200.1 | 7 | 1198 | |||
XM_011532899.2 | XP_011531201.1 | 7 | 1199 | |||
XM_011532901.2 | XP_011531203.1 | 7 | 1199 | |||
XM_017004258.1 | XP_016859747.1 | 6 | 955 | |||
XM_017004259.1 | XP_016859748.1 | 6 | 1216 | |||
XM_017004260.1 | XP_016859749.1 | 5 | 830 | |||
XM_017004261.1 | XP_016859750.1 | 5 | 490 | |||
XM_017004262.1 | XP_016859751.1 | 4 | 373 | |||
GenBank mRNA | AB209139.1 | - | 6 | 1077 | ||
AF167706.1 | - | 6 | 1050 | |||
AY358371.1 | - | 6 | 1378 | |||
BC111989.1 | - | 6 | 1048 | 8327457 | ||
BC113371.1 | - | 6 | 1048 | 8322423 |