Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016445.2 | NP_057529.1 | 9 | 1412 | ||
XM_011536644.2 | XP_011534946.1 | 8 | 1284 | |||
GenBank mRNA | AF228603.1 | - | 9 | 1323 | ||
BC001226.1 | - | 9 | 1356 | 3457876 | ||
BC008056.2 | - | 9 | 1266 | |||
BX114063.1 | - | 4 | 542 |