Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204090.1 | NP_001191019.1 | 3 | 619 | ||
NM_016468.6 | NP_057552.1 | 4 | 691 | |||
GenBank mRNA | AF151037.1 | - | 4 | 639 | ||
AF226729.1 | - | 4 | 665 | |||
BC001702.2 | - | 4 | 592 | 2966935 | ||
CB156527.1 | - | 3 | 563 | |||
FJ460510.1 | - | 4 | 691 | |||
FJ460511.1 | - | 4 | 691 |
Gene Symbol: SYNJ2BP-COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202547.2 | NP_001189476.1 | 6 | 929 | ||
NM_001202548.2 | NP_001189477.1 | 6 | 902 | |||
NM_001202549.2 | NP_001189478.1 | 5 | 830 |