Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286371.1 | NP_001273300.1 | 6 | 791 | ||
NM_001286372.1 | NP_001273301.1 | 5 | 696 | |||
NM_016485.4 | NP_057569.2 | 6 | 791 | |||
GenBank mRNA | AF060225.1 | - | 6 | 681 | ||
AF151062.1 | - | 6 | 688 | |||
AF271994.1 | - | 6 | 680 | |||
AJ420507.1 | - | 6 | 686 | |||
AK000051.1 | - | 6 | 680 | |||
AK130738.1 | - | 6 | 683 | |||
AK301376.1 | - | 5 | 599 | |||
AL136684.1 | - | 6 | 684 | |||
AY007093.1 | - | 5 | 495 | |||
BC005937.1 | - | 6 | 676 | 4081455 | ||
BC006989.1 | - | 6 | 690 | 3961637 | ||
BC013217.1 | - | 2 | 184 | |||
BC022536.1 | - | 6 | 690 | 4817057 | ||
BM806661.1 | - | 6 | 591 | |||
CR457207.1 | - | 6 | 665 |