Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016485.4 | NP_057569.2 | 6, 7 | 831 | ||
GenBank mRNA | AF060225.1 | - | 6, 7 | 721 | ||
AF141341.1 | - | 6, 7 | 709 | |||
AF151062.1 | - | 6, 7 | 728 | |||
AF271994.1 | - | 6, 7 | 720 | |||
AJ420507.1 | - | 6, 7 | 726 | |||
AK000051.1 | - | 6, 7 | 720 | |||
AK130738.1 | - | 6, 7 | 723 | |||
AL136684.1 | - | 6, 7 | 724 | |||
AY007093.1 | - | 5, 6 | 535 | |||
BC005937.1 | - | 6, 7 | 716 | 4081455 | ||
BC006989.1 | - | 6, 7 | 730 | 3961637 | ||
BC013217.1 | - | 2, 3 | 224 | |||
BC022536.1 | - | 6, 7 | 730 | 4817057 | ||
CR457207.1 | - | 6, 7 | 705 |