Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131027.1 | NP_001124499.1 | 3 | 387 | ||
NM_003687.3 | NP_003678.2 | 3 | 387 | |||
GenBank mRNA | AF153882.1 | - | 3 | 351 | ||
AF154335.1 | - | 3 | 374 | |||
AK225407.1 | - | 3 | 349 | |||
AK313508.1 | - | 3 | 352 | |||
BC003096.1 | - | 3 | 338 | 3501794 | ||
BC016765.1 | - | 3 | 389 | 4132234 | ||
BT007019.1 | - | 3 | 310 | |||
DN997097.1 | - | 3 | 406 | |||
KU178433.1 | - | 3 | 310 | |||
KU178434.1 | - | 3 | 310 | |||
X93510.1 | - | 3 | 348 |