Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPHLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143787.1 | NP_001137259.1 | 5 | 489 | ||
NM_016488.6 | NP_057572.5 | 6 | 654 | |||
NM_201438.1 | NP_958846.1 | 6 | 532 | |||
NM_201439.1 | NP_958847.1 | 6 | 654 | |||
NM_201440.1 | NP_958848.1 | 6 | 532 | |||
NM_201515.1 | NP_958923.1 | 7 | 697 | |||
XM_011538459.2 | XP_011536761.1 | 7 | 675 | |||
XM_011538467.2 | XP_011536769.1 | 6 | 514 | |||
XM_017019434.1 | XP_016874923.1 | 7 | 675 | |||
XM_017019435.1 | XP_016874924.1 | 7 | 674 | |||
XM_017019436.1 | XP_016874925.1 | 7 | 675 | |||
XM_017019437.1 | XP_016874926.1 | 6 | 632 | |||
XM_017019438.1 | XP_016874927.1 | 6 | 632 | |||
XM_017019439.1 | XP_016874928.1 | 7 | 675 | |||
XM_017019444.1 | XP_016874933.1 | 6 | 514 | |||
XM_017019445.1 | XP_016874934.1 | 5 | 508 | |||
XM_017019446.1 | XP_016874935.1 | 5 | 507 | |||
GenBank mRNA | AF151040.1 | - | 6 | 495 | ||
AK055690.1 | - | 6 | 642 | |||
AK129910.1 | - | 7 | 697 | |||
AK303612.1 | - | 5 | 489 | |||
AY039238.1 | - | 6 | 636 | |||
AY157850.1 | - | 7 | 693 | |||
BC025306.1 | - | 5 | 456 | |||
BC039832.1 | - | 6 | 499 | 6051282 | ||
BC040449.1 | - | 6 | 499 | |||
CF242917.1 | - | 5 | 504 |