Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171201.1 | NP_001164672.1 | 2 | 304 | ||
NM_001171203.2 | NP_001164674.1 | 4 | 500 | |||
NM_001171204.2 | NP_001164675.1 | 4 | 468 | |||
NM_016525.4 | NP_057609.2 | 3 | 345 | |||
NR_033243.2 | 4 | 461 | ||||
XM_006716779.3 | XP_006716842.1 | 3 | 371 | |||
XM_011517899.1 | XP_011516201.1 | 4 | 402 | |||
XM_017014801.1 | XP_016870290.1 | 4 | 737 | |||
GenBank mRNA | AF222043.2 | - | 3 | 274 | ||
AK074426.1 | - | 3 | 338 | |||
AK074724.1 | - | 1 | 83 | |||
AK074745.1 | - | 3 | 305 | |||
AK074812.1 | - | 3 | 345 | |||
AK074969.1 | - | 4 | 447 | |||
AK074995.1 | - | 3 | 309 | |||
AK223235.1 | - | 3 | 300 | |||
AK303711.1 | - | 2 | 304 | |||
AL136733.1 | - | 3 | 138 | |||
BC020950.1 | - | 3 | 307 | 3851813 | ||
BC098141.1 | - | 3 | 241 | 40011247 | ||
BC098316.1 | - | 3 | 241 | 40011248 | ||
BC099726.1 | - | 3 | 241 | 40011249 | ||
BC100668.1 | - | 3 | 241 | 40011243 | ||
BP237024.1 | - | 4 | 429 | |||
BX473128.1 | - | 4 | 453 | |||
CR533551.1 | - | 2 | 102 | |||
DA741347.1 | - | 4 | 455 |