Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACKR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016557.3 | NP_057641.1 | 2 | 453 | ||
NM_178445.2 | NP_848540.1 | 1 | 618 | |||
GenBank mRNA | AF110640.1 | - | 2 | 397 | ||
AK225927.1 | - | 1 | 367 | |||
AK304461.1 | - | 2 | 453 | |||
AK313923.1 | - | 2 | 420 | |||
BC069438.1 | - | 2 | 397 | 7262320 | ||
BC095501.1 | - | 1 | 354 | |||
DC419249.1 | - | 2 | 454 |