Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016561.2 | NP_057645.1 | 4 | 786 | ||
XM_005255350.2 | XP_005255407.1 | 3 | 561 | |||
GenBank mRNA | AF173003.1 | - | 4 | 658 | ||
AK291114.1 | - | 4 | 594 | |||
AK300778.1 | - | 2 | 359 | |||
AK313890.1 | - | 3 | 594 | |||
BC003054.1 | - | 4 | 617 | 3537783 |