Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016584.2 | NP_057668.1 | 4 | 825 | ||
XM_011538477.2 | XP_011536779.1 | 6 | 1012 | |||
GenBank mRNA | AB030000.1 | - | 4 | 802 | ||
AF301620.1 | - | 4 | 809 | |||
AY359083.1 | - | 4 | 829 |