Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135179.1 | NP_001128651.1 | 7 | 1648 | ||
NM_016598.2 | NP_057682.1 | 8 | 1732 | |||
XM_006713192.2 | XP_006713255.1 | 9 | 1834 | |||
XM_006713193.2 | XP_006713256.1 | 8 | 1751 | |||
XM_011533801.2 | XP_011532103.1 | 7 | 1619 | |||
XM_017006566.1 | XP_016862055.1 | 8 | 1686 | |||
XM_017006567.1 | XP_016862056.1 | 8 | 1724 | |||
XM_017006569.1 | XP_016862058.1 | 8 | 1723 | |||
XM_017006571.1 | XP_016862060.1 | 8 | 1728 | |||
GenBank mRNA | AF052182.1 | - | 1 | 260 | ||
AF247703.1 | - | 8 | 1672 | |||
BC020350.1 | - | 1 | 426 | |||
BC020424.1 | - | 1 | 426 | |||
BC038436.1 | - | 1 | 426 |