Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135647.1 | NP_001129119.1 | 5 | 2103 | ||
NM_016605.2 | NP_057689.1 | 5 | 2078 | |||
GenBank mRNA | AF251040.1 | - | 5 | 1814 | ||
AK027412.1 | - | 3 | 969 | |||
BC000259.1 | - | 5 | 1820 | 3357346 |