Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135647.1 | NP_001129119.1 | 5 | 3966 | ||
NM_016605.2 | NP_057689.1 | 5 | 3941 | |||
GenBank mRNA | AF251040.1 | - | 5 | 3679 | ||
AK027412.1 | - | 3 | 2834 | |||
BC000259.1 | - | 6 | 2145 | 3357346 |