Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271803.1 | NP_001258732.1 | 8 | 1303 | ||
NM_016606.3 | NP_057690.2 | 8 | 1297 | |||
NR_073448.1 | 8 | 1372 | ||||
NR_073449.1 | 8 | 1378 | ||||
GenBank mRNA | AF070551.1 | - | 4 | 741 | ||
AF251041.1 | - | 7 | 1077 | |||
AK056193.1 | - | 8 | 1259 | |||
AK223611.1 | - | 8 | 1244 | |||
BC006218.2 | - | 8 | 1211 | 3940709 | ||
BC148256.1 | - | 1 | 428 |