Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016608.1 | NP_057692.1 | 4 | 481 | ||
GenBank mRNA | AB039670.1 | - | 4 | 481 | ||
AF248963.1 | - | 4 | 414 | |||
AK074785.1 | - | 4 | 431 | |||
AK222578.1 | - | 4 | 458 | |||
BC002691.2 | - | 4 | 420 | 3605690 |