Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016622.3 | NP_057706.2 | 4 | 485 | ||
NM_145644.2 | NP_663619.1 | 4 | 485 | |||
XM_005264361.2 | XP_005264418.1 | 4 | 563 | |||
GenBank mRNA | AF208849.1 | - | 4 | 445 | ||
AK074352.1 | - | 4 | 457 | |||
AK314555.1 | - | 4 | 455 | |||
AK314577.1 | - | 4 | 455 | |||
AL832982.1 | - | 4 | 433 | |||
BC020651.1 | - | 4 | 461 | 4709909 | ||
BQ421154.1 | - | 3 | 322 | |||
DA443416.1 | - | 4 | 461 |