Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016622.3 | NP_057706.2 | 4 | 2704 | ||
NM_145644.2 | NP_663619.1 | 5 | 892 | |||
GenBank mRNA | AK026220.1 | - | 1 | 1307 | ||
AK074352.1 | - | 4 | 2676 | |||
AL832982.1 | - | 4 | 2652 | |||
BC020651.1 | - | 5 | 868 | 4709909 | ||
BU195549.1 | - | 1 | 161 |