Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320108.1 | NP_001307037.1 | 1 | 2214 | ||
NM_003708.4 | NP_003699.3 | 1 | 2214 | |||
GenBank mRNA | AB209332.1 | - | 1 | 2214 | ||
AF057034.1 | - | 1 | 438 | |||
AF086735.1 | - | 1 | 1154 | |||
AK311025.1 | - | 3 | 1052 |