Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114086.1 | NP_001107558.1 | 5 | 1009 | ||
NM_001256023.1 | NP_001242952.1 | 5 | 773 | |||
NM_016929.4 | NP_058625.2 | 5 | 773 | |||
NR_045672.1 | 2 | 351 | ||||
NR_045673.1 | 3 | 390 | ||||
NR_045674.1 | 4 | 724 | ||||
XM_011514692.2 | XP_011512994.1 | 5 | 1009 | |||
XM_011514694.2 | XP_011512996.1 | 5 | 1147 | |||
XM_017010953.1 | XP_016866442.1 | 6 | 1254 | |||
GenBank mRNA | AF176920.1 | - | 5 | 573 | ||
AF216941.1 | - | 5 | 723 | |||
AK075144.1 | - | 5 | 712 | |||
AK075163.1 | - | 5 | 624 | |||
AK097048.1 | - | 5 | 1009 | |||
AK222544.1 | - | 5 | 741 | |||
AK223130.1 | - | 5 | 744 | |||
AK314416.1 | - | 5 | 589 | |||
AY032602.1 | - | 5 | 903 | |||
BC035968.1 | - | 5 | 741 | 4611102 | ||
BC039380.1 | - | 4 | 728 | |||
BI559571.1 | - | 3 | 342 | |||
BP369816.1 | - | 2 | 296 | |||
DQ679794.1 | - | 5 | 741 |