Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079514.2 | NP_001072982.1 | 4, 5 | 1056 | ||
NM_001288656.1 | NP_001275585.1 | 3, 4 | 490 | |||
XM_005255277.3 | XP_005255334.1 | 4, 5 | 1241 | |||
XM_005255278.3 | XP_005255335.1 | 4, 5 | 2205 | |||
XM_005255279.3 | XP_005255336.1 | 4, 5 | 1241 | |||
XM_011522465.1 | XP_011520767.1 | 4, 5 | 1241 | |||
XM_011522467.1 | XP_011520769.1 | 4, 5 | 1241 | |||
XM_017023189.1 | XP_016878678.1 | 4, 5 | 1241 | |||
XM_017023190.1 | XP_016878679.1 | 4, 5 | 1241 | |||
XM_017023192.1 | XP_016878681.1 | 4, 5 | 1241 | |||
GenBank mRNA | AF108460.1 | - | 4, 5 | 551 | ||
AF108461.1 | - | 3, 4 | 1138 | |||
AK056282.1 | - | 4, 5 | 2205 | |||
AK300131.1 | - | 3, 4 | 490 | |||
AW385352.1 | - | 1, 2 | 69 | |||
BC013750.1 | - | 4, 5 | 776 |