Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017419.2 | NP_059115.1 | 8, 9 | 1275 | ||
XM_017008291.1 | XP_016863780.1 | 7, 8 | 2179 | |||
GenBank mRNA | AJ252011.1 | - | 8, 9 | 1275 |