Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098525.2 | NP_001091995.1 | 4 | 700 | ||
NM_001286686.1 | NP_001273615.1 | 4 | 638 | |||
NM_001286687.1 | NP_001273616.1 | 4 | 697 | |||
NM_018204.4 | NP_060674.3 | 4 | 697 | |||
XM_005266344.4 | XP_005266401.1 | 4 | 608 | |||
XM_011535043.2 | XP_011533345.1 | 3 | 552 | |||
GenBank mRNA | AF177227.1 | - | 3 | 392 | ||
AK022982.1 | - | 4 | 596 | |||
AK299083.1 | - | 4 | 638 | |||
AL136848.1 | - | 1 | 102 | |||
AY062261.1 | - | 4 | 618 | |||
AY062262.1 | - | 4 | 618 | |||
BC130296.1 | - | 4 | 545 | 40146327 | ||
BC136332.1 | - | 4 | 545 | 9020319 | ||
BM541375.1 | - | 4 | 702 | |||
EF560732.1 | - | 4 | 581 | |||
Y15758.1 | - | 4 | 593 |