Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098525.2 | NP_001091995.1 | 9 | 2602 | ||
NM_001286686.1 | NP_001273615.1 | 9 | 2540 | |||
NM_018204.4 | NP_060674.3 | 9 | 2599 | |||
XM_005266344.4 | XP_005266401.1 | 9 | 2510 | |||
XM_011535043.2 | XP_011533345.1 | 8 | 2454 | |||
GenBank mRNA | AK001611.1 | - | 6 | 1475 | ||
AK022982.1 | - | 9 | 2498 | |||
AK027103.1 | - | 2 | 646 | |||
AL136848.1 | - | 6 | 2004 | |||
AY062261.1 | - | 9 | 2520 | |||
BC130296.1 | - | 9 | 2447 | 40146327 | ||
BC136332.1 | - | 9 | 2447 | 9020319 | ||
Y15758.1 | - | 9 | 2495 |