Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018945.3 | NP_061818.1 | 13 | 1857 | ||
XM_005266931.3 | XP_005266988.1 | 12 | 4706 | |||
XM_011535755.2 | XP_011534057.1 | 14 | 1686 | |||
XM_017010741.1 | XP_016866230.1 | 12 | 2492 | |||
GenBank mRNA | AB038040.1 | - | 13 | 1859 | ||
AB209990.1 | - | 12 | 1963 | |||
AJ251860.1 | - | 13 | 1851 | |||
AK290582.1 | - | 13 | 1890 |