Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129981.2 | NP_001123453.1 | 8 | 1375 | ||
NM_001291218.1 | NP_001278147.1 | 9 | 1474 | |||
NM_001291219.1 | NP_001278148.1 | 8 | 1375 | |||
NM_020349.3 | NP_065082.2 | 9 | 1474 | |||
GenBank mRNA | AB058599.1 | - | 9 | 1136 | ||
AJ304805.1 | - | 9 | 1125 | |||
AJ346356.1 | - | 3 | 410 | |||
AJ583444.1 | - | 9 | 1463 | |||
AK056990.1 | - | 9 | 1416 | |||
BC020817.1 | - | 8 | 1037 | |||
BC107759.1 | - | 9 | 1377 | 4096546 |