Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 6 | 1994 | ||
XM_005255254.3 | XP_005255311.1 | 6 | 1984 | |||
XM_005255257.4 | XP_005255314.1 | 5 | 1961 | |||
XM_017023143.1 | XP_016878632.1 | 5 | 1938 | |||
XM_017023144.1 | XP_016878633.1 | 5 | 1938 | |||
XM_017023145.1 | XP_016878634.1 | 5 | 1940 | |||
XM_017023146.1 | XP_016878635.1 | 5 | 2003 | |||
XM_017023147.1 | XP_016878636.1 | 6 | 2178 | |||
XM_017023148.1 | XP_016878637.1 | 5 | 1935 | |||
XM_017023149.1 | XP_016878638.1 | 5 | 1935 | |||
XM_017023150.1 | XP_016878639.1 | 5 | 1936 | |||
XM_017023151.1 | XP_016878640.1 | 5 | 1933 | |||
XM_017023152.1 | XP_016878641.1 | 4 | 1510 | |||
XM_017023153.1 | XP_016878642.1 | 5 | 1961 | |||
XM_017023154.1 | XP_016878643.1 | 4 | 1885 | |||
GenBank mRNA | AB040893.1 | - | 1 | 194 | ||
AJ492221.1 | - | 5 | 1880 | |||
AY035864.1 | - | 3 | 1528 |