Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 23, 24 | 5483 | ||
XM_005255254.3 | XP_005255311.1 | 22, 23 | 5326 | |||
XM_005255257.4 | XP_005255314.1 | 22, 23 | 5450 | |||
XM_017023143.1 | XP_016878632.1 | 22, 23 | 5427 | |||
XM_017023144.1 | XP_016878633.1 | 21, 22 | 5394 | |||
XM_017023145.1 | XP_016878634.1 | 22, 23 | 5360 | |||
XM_017023146.1 | XP_016878635.1 | 22, 23 | 5492 | |||
XM_017023147.1 | XP_016878636.1 | 22, 23 | 5520 | |||
XM_017023148.1 | XP_016878637.1 | 21, 22 | 5247 | |||
XM_017023149.1 | XP_016878638.1 | 20, 21 | 5244 | |||
XM_017023150.1 | XP_016878639.1 | 21, 22 | 5209 | |||
XM_017023151.1 | XP_016878640.1 | 20, 21 | 5098 | |||
XM_017023152.1 | XP_016878641.1 | 21, 22 | 4999 | |||
XM_017023153.1 | XP_016878642.1 | 22, 23 | 5450 | |||
XM_017023154.1 | XP_016878643.1 | 20, 21 | 5341 | |||
GenBank mRNA | AB040893.1 | - | 18, 19 | 3683 | ||
AJ492221.1 | - | 22, 23 | 5369 | |||
AK001773.1 | - | 6, 7 | 808 | |||
AL833757.1 | - | 17, 18 | 2644 | |||
AY035864.1 | - | 20, 21 | 5017 | |||
BC024324.1 | - | 6, 7 | 829 | |||
BC068209.1 | - | 1, 2 | 354 |