Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281734.1 | NP_001268663.1 | 12 | 3575 | ||
NM_001281735.1 | NP_001268664.1 | 9 | 1753 | |||
NM_021260.3 | NP_067083.1 | 12 | 3617 | |||
NM_178441.2 | NP_848535.1 | 9 | 1758 | |||
XM_017021373.1 | XP_016876862.1 | 10 | 2282 | |||
GenBank mRNA | AB046809.1 | - | 12 | 3442 | ||
AF318319.1 | - | 9 | 1786 | |||
AJ310569.1 | - | 12 | 3468 | |||
BC014902.1 | - | 12 | 3494 | |||
BC053520.1 | - | 12 | 3545 | 5493027 | ||
CK005173.1 | - | 1 | 334 |