Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022044.2 | NP_071327.2 | 1 | 260 | ||
GenBank mRNA | AB043007.1 | - | 1 | 196 | ||
AF277316.1 | - | 1 | 184 | |||
AY359118.1 | - | 1 | 227 | |||
BC132849.1 | - | 1 | 252 | 40146871 | ||
BC132851.1 | - | 1 | 252 | 40146873 | ||
CR456570.1 | - | 1 | 185 |