Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003736.2 | NP_003727.1 | 1 | 751 | ||
NM_032098.1 | NP_115269.1 | 1 | 751 | |||
GenBank mRNA | AF152333.1 | - | 1 | 751 | ||
AF152520.1 | - | 1 | 751 | |||
BC112088.1 | - | 1 | 798 | 8327556 | ||
BC113607.1 | - | 1 | 800 | 8322659 |