Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318014.1 | NP_001304943.1 | 4 | 2624 | ||
NM_001318022.1 | NP_001304951.1 | 4 | 2507 | |||
NM_030883.4 | NP_112145.1 | 4 | 2511 | |||
XM_017010739.1 | XP_016866228.1 | 5 | 2996 | |||
GenBank mRNA | AJ459849.1 | - | 1 | 37 | ||
AK127984.1 | - | 3 | 1710 | |||
BC048991.2 | - | 3 | 2459 | 5271584 |