Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 14 | 2094 | ||
XM_011518446.2 | XP_011516748.1 | 14 | 2109 | |||
XM_011518447.1 | XP_011516749.1 | 13 | 2070 | |||
XM_011518449.1 | XP_011516751.1 | 13 | 1977 | |||
XM_011518451.1 | XP_011516753.1 | 12 | 1965 | |||
XM_011518452.1 | XP_011516754.1 | 13 | 1860 | |||
XM_017014571.1 | XP_016870060.1 | 13 | 2067 | |||
XM_017014572.1 | XP_016870061.1 | 12 | 1836 | |||
GenBank mRNA | AF153415.1 | - | 14 | 2094 | ||
AK074248.1 | - | 4 | 392 | |||
AK172815.1 | - | 8 | 839 | |||
AK299952.1 | - | 14 | 2076 | |||
AL556162.3 | - | 8 | 805 | |||
AL833960.1 | - | 12 | 1485 | |||
BC014248.2 | - | 4 | 445 |