Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 16 | 2365 | ||
XM_011518446.2 | XP_011516748.1 | 16 | 2380 | |||
XM_011518447.1 | XP_011516749.1 | 15 | 2341 | |||
XM_011518449.1 | XP_011516751.1 | 15 | 2248 | |||
XM_011518451.1 | XP_011516753.1 | 14 | 2236 | |||
XM_011518452.1 | XP_011516754.1 | 15 | 2131 | |||
XM_017014571.1 | XP_016870060.1 | 15 | 2338 | |||
XM_017014572.1 | XP_016870061.1 | 14 | 2107 | |||
GenBank mRNA | AF153415.1 | - | 16 | 2365 | ||
AK074248.1 | - | 6 | 663 | |||
AK090543.1 | - | 2 | 1420 | |||
AK172815.1 | - | 10 | 1110 | |||
AK299952.1 | - | 16 | 2347 | |||
AL833960.1 | - | 14 | 1756 | |||
BC014248.2 | - | 6 | 716 |