Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032554.3 | NP_115943.1 | 1 | 2790 | ||
GenBank mRNA | AF385431.1 | - | 1 | 2790 |