Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNISR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322405.1 | NP_001309334.1 | 11 | 1548 | ||
NM_001322406.1 | NP_001309335.1 | 12 | 1664 | |||
NM_001322408.1 | NP_001309337.1 | 10 | 1468 | |||
NM_001322410.1 | NP_001309339.1 | 11 | 1552 | |||
NM_001322412.1 | NP_001309341.1 | 11 | 1548 | |||
NM_001322413.1 | NP_001309342.1 | 10 | 1472 | |||
NM_001322414.1 | NP_001309343.1 | 10 | 1498 | |||
NM_001322415.1 | NP_001309344.1 | 9 | 1418 | |||
NM_001322416.1 | NP_001309345.1 | 11 | 1552 | |||
NM_015491.2 | NP_056306.1 | 10 | 1472 | |||
NM_032870.3 | NP_116259.2 | 11 | 1552 | |||
NR_136326.1 | 9 | 2192 | ||||
XM_005266912.3 | XP_005266969.2 | 10 | 1431 | |||
XM_017010710.1 | XP_016866199.1 | 10 | 1495 | |||
XM_017010711.1 | XP_016866200.1 | 9 | 1381 | |||
XM_017010712.1 | XP_016866201.1 | 11 | 2903 | |||
XM_017010713.1 | XP_016866202.1 | 11 | 2899 | |||
GenBank mRNA | AF161424.1 | - | 5 | 599 | ||
AF314184.1 | - | 9 | 1342 | |||
AF314186.1 | - | 11 | 1490 | |||
AK027658.1 | - | 10 | 2235 | |||
AK027898.1 | - | 11 | 1508 | |||
AK074628.1 | - | 10 | 1435 | |||
BC012780.2 | - | 11 | 1494 | |||
BC064413.1 | - | 11 | 1487 |