Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163499.1 | NP_001156971.1 | 8 | 1184 | ||
NM_012172.4 | NP_036304.2 | 8 | 1334 | |||
NM_033506.2 | NP_277041.1 | 8 | 1445 | |||
XM_005250259.3 | XP_005250316.1 | 8 | 1619 | |||
XM_011516022.1 | XP_011514324.1 | 8 | 1177 | |||
XM_011516023.2 | XP_011514325.1 | 7 | 1436 | |||
XM_017011961.1 | XP_016867450.1 | 9 | 2920 | |||
GenBank mRNA | AK292240.1 | - | 8 | 1452 | ||
AK301864.1 | - | 8 | 1278 | |||
AK302252.1 | - | 8 | 1184 | |||
AL136811.1 | - | 8 | 1390 | |||
BC067293.1 | - | 8 | 1385 | 30383439 |