Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080817.4 | NP_543007.1 | 3 | 595 | ||
XM_017029414.1 | XP_016884903.1 | 2 | 751 | |||
GenBank mRNA | AJ319076.1 | - | 1 | 373 | ||
AK122626.1 | - | 3 | 569 | |||
AK313702.1 | - | 3 | 570 | |||
AL832460.1 | - | 3 | 570 | |||
BC070180.1 | - | 3 | 595 | 30351329 |
Set Membership: |
G-protein coupled receptor |