Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003747.2 | NP_003738.2 | 1, 2 | 686 | ||
XM_006716263.3 | XP_006716326.1 | 1, 2 | 1392 | |||
XM_011543845.2 | XP_011542147.1 | 1, 2 | 1392 | |||
XM_011543846.2 | XP_011542148.1 | 1, 2 | 1392 | |||
XM_011543847.2 | XP_011542149.1 | 1, 2 | 1391 | |||
GenBank mRNA | AF082556.1 | - | 1, 2 | 686 | ||
AK301798.1 | - | 1, 2 | 607 | |||
AK302031.1 | - | 1, 2 | 707 | |||
BC098394.1 | - | 1, 2 | 709 | 4374347 |