Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139265.3 | NP_644670.1 | 2 | 411 | ||
XM_017022100.1 | XP_016877589.1 | 2 | 278 | |||
GenBank mRNA | AF181265.1 | - | 2 | 171 | ||
AF307137.1 | - | 2 | 399 | |||
AF323924.1 | - | 2 | 407 | |||
AF454953.1 | - | 2 | 398 | |||
AK074621.1 | - | 2 | 406 | |||
AK292634.1 | - | 2 | 408 | |||
BC006287.2 | - | 2 | 398 | 3958645 | ||
BC051823.1 | - | 2 | 402 | 4944876 | ||
BP314075.1 | - | 2 | 411 |