Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139265.3 | NP_644670.1 | 6 | 2542 | ||
XM_017022100.1 | XP_016877589.1 | 6 | 2409 | |||
GenBank mRNA | AF086163.1 | - | 2 | 219 | ||
AF181265.1 | - | 6 | 2300 | |||
AF454953.1 | - | 6 | 2528 | |||
AK024902.1 | - | 4 | 1732 | |||
BC006287.2 | - | 6 | 2529 | 3958645 | ||
BC051823.1 | - | 6 | 2533 | 4944876 |