Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 6 | 687 | ||
NM_024021.3 | NP_076926.2 | 8 | 992 | |||
NM_148975.2 | NP_683876.1 | 7 | 846 | |||
XM_017017909.1 | XP_016873398.1 | 7 | 859 | |||
GenBank mRNA | AB013102.1 | - | 7 | 687 | ||
AF068288.1 | - | 7 | 780 | |||
AF237912.1 | - | 7 | 797 | |||
AF350500.1 | - | 6 | 750 | |||
AF354928.1 | - | 5 | 511 | |||
AK075081.1 | - | 8 | 877 | |||
AK292089.1 | - | 7 | 801 | |||
BC020648.1 | - | 7 | 721 | 4720088 | ||
CB046393.1 | - | 6 | 556 |