Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291488.1 | NP_001278417.1 | 4, 5 | 245 | ||
NM_001291489.1 | NP_001278418.1 | 3, 4 | 315 | |||
NM_152354.5 | NP_689567.4 | 3, 4 | 224 | |||
GenBank mRNA | AL833447.1 | - | 3, 4 | 202 | ||
AY166790.1 | - | 1, 2 | 14 | |||
BC117300.1 | - | 2, 3 | 188 | 40125851 | ||
BC117302.1 | - | 2, 3 | 188 | 40125853 | ||
BC143984.1 | - | 3, 4 | 209 | |||
DA793212.1 | - | 3, 4 | 288 | |||
DA949487.1 | - | 4, 5 | 245 |