Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291488.1 | NP_001278417.1 | 5 | 263 | ||
NM_001291489.1 | NP_001278418.1 | 4 | 333 | |||
NM_001291490.1 | NP_001278419.1 | 5 | 397 | |||
NM_001291491.1 | NP_001278420.1 | 3 | 115 | |||
NM_152354.5 | NP_689567.4 | 4 | 242 | |||
GenBank mRNA | AL833447.1 | - | 4 | 220 | ||
AY166790.1 | - | 2 | 32 | |||
BC117300.1 | - | 3 | 206 | 40125851 | ||
BC117302.1 | - | 3 | 206 | 40125853 | ||
BC143984.1 | - | 4 | 227 | |||
DA202784.1 | - | 3 | 112 | |||
DA793212.1 | - | 4 | 306 | |||
DA949487.1 | - | 5 | 263 |