Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037283.1 | NP_001032360.1 | 1, 2 | 586 | ||
NM_003751.3 | NP_003742.2 | 1, 2 | 586 | |||
XM_011515599.1 | XP_011513901.1 | 1, 2 | 885 | |||
XM_011515600.1 | XP_011513902.1 | 1, 2 | 885 | |||
XM_011515601.1 | XP_011513903.1 | 2, 3 | 768 | |||
XM_017012752.1 | XP_016868241.1 | 1, 2 | 885 | |||
XM_017012753.1 | XP_016868242.1 | 1, 2 | 885 | |||
GenBank mRNA | AK300967.1 | - | 2, 3 | 328 | ||
AK302058.1 | - | 2, 3 | 112 | |||
BC001173.1 | - | 1, 2 | 534 | 3501759 | ||
BC110865.1 | - | 1, 2 | 512 | 6175284 | ||
BG325094.1 | - | 1, 2 | 555 | |||
BM556863.1 | - | 349 | ||||
BQ070291.1 | - | 432 | ||||
U62583.1 | - | 1, 2 | 556 | |||
U78525.1 | - | 1, 2 | 578 |