Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139488.1 | NP_001132960.1 | 18 | 3519 | ||
NM_015376.2 | NP_056191.1 | 17 | 3074 | |||
NM_170672.2 | NP_733772.1 | 19 | 3432 | |||
XM_005264246.3 | XP_005264303.1 | 18 | 3856 | |||
XM_011532746.2 | XP_011531048.1 | 19 | 3306 | |||
XM_011532747.2 | XP_011531049.1 | 18 | 3321 | |||
XM_011532748.2 | XP_011531050.2 | 18 | 3167 | |||
XM_017003759.1 | XP_016859248.1 | 20 | 4782 | |||
XM_017003760.1 | XP_016859249.1 | 19 | 5254 | |||
XM_017003761.1 | XP_016859250.1 | 18 | 3184 | |||
XM_017003762.1 | XP_016859251.1 | 19 | 3430 | |||
XM_017003763.1 | XP_016859252.1 | 18 | 3160 | |||
GenBank mRNA | AB020653.1 | - | 17 | 3068 | ||
BC027849.1 | - | 19 | 3428 | 5203041 | ||
BX114300.1 | - | 1 | 626 | |||
BX647990.1 | - | 18 | 3116 |