Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012093.3 | NP_036225.2 | 3 | 1325 | ||
NM_174858.2 | NP_777283.1 | 3 | 714 | |||
XM_005270739.4 | XP_005270796.1 | 3 | 561 | |||
XM_006710572.3 | XP_006710635.1 | 3 | 492 | |||
XM_017001008.1 | XP_016856497.1 | 3 | 509 | |||
XM_017001009.1 | XP_016856498.1 | 3 | 1217 | |||
XM_017001010.1 | XP_016856499.1 | 3 | 1544 | |||
XM_017001011.1 | XP_016856500.1 | 3 | 327 | |||
XM_017001012.1 | XP_016856501.1 | 3 | 783 | |||
GenBank mRNA | AF445193.1 | - | 3 | 714 | ||
AK090967.1 | - | 3 | 561 | |||
AY171600.2 | - | 3 | 1330 | |||
BC033896.1 | - | 3 | 1330 | 4818069 | ||
BC036666.1 | - | 3 | 640 | 4816351 |